Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igf1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010513.2 | NP_034643.2 | 2 | 733 | ||
GenBank mRNA | AF056187.1 | - | 2 | 131 | ||
BC138868.1 | - | 2 | 163 | 8861890 | ||
BC138869.1 | - | 2 | 163 | 8861891 | ||
U00182.1 | - | 2 | 133 |