Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igf1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010513.2 | NP_034643.2 | 12 | 3190 | ||
GenBank mRNA | AF056187.1 | - | 12 | 2588 | ||
AK137220.1 | - | 7 | 1319 | |||
AK155884.1 | - | 11 | 2086 | |||
AK162325.1 | - | 1 | 404 | |||
BC138868.1 | - | 12 | 2620 | 8861890 | ||
BC138869.1 | - | 12 | 2620 | 8861891 |