Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igfbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010517.3 | NP_034647.1 | 4 | 1250 | ||
GenBank mRNA | AK003243.1 | - | 1 | 254 | ||
AK031212.1 | - | 4 | 1267 | |||
AK150229.1 | - | 4 | 1263 | |||
AK165688.1 | - | 1268 | ||||
AK197851.1 | - | 1 | 376 | |||
AK198090.1 | - | 1 | 377 | |||
AK200478.1 | - | 1 | 314 | |||
AK200553.1 | - | 1 | 375 | |||
BC019836.1 | - | 4 | 1250 | 5123738 |