Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207053.1 | NP_001193982.1 | 4 | 683 | ||
NM_001243126.1 | NP_001230055.1 | 4 | 666 | |||
NM_005046.3 | NP_005037.1 | 5 | 814 | |||
NM_139277.2 | NP_644806.1 | 5 | 644 | |||
GenBank mRNA | AF411214.1 | - | 5 | 788 | ||
AF411215.1 | - | 4 | 657 | |||
AK289660.1 | - | 5 | 667 | |||
AK295313.1 | - | 4 | 666 | |||
AY601109.1 | - | 4 | 542 | |||
AY646152.1 | - | 3 | 398 | |||
BC032005.1 | - | 5 | 653 | 4750452 | ||
DQ018784.1 | - | 4 | 542 | |||
KF963189.1 | - | 4 | 506 | |||
KF963190.1 | - | 4 | 507 | |||
KF963191.1 | - | 4 | 507 | |||
KF963192.1 | - | 3 | 101 | |||
KF963193.1 | - | 4 | 507 | |||
L33404.1 | - | 4 | 557 |