Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010551.3 | NP_034681.2 | 14 | 3047 | ||
GenBank mRNA | AF006001.1 | - | 1 | 777 | ||
AF017111.1 | - | 2 | 927 | |||
AF175292.1 | - | 14 | 3047 | |||
AK038319.1 | - | 2 | 963 | |||
AK083338.1 | - | 2 | 856 | |||
AK134007.1 | - | 2 | 878 | |||
AK137802.1 | - | 2 | 934 | |||
AK169551.1 | - | 2 | 950 | |||
AK220455.1 | - | 14 | 3057 | |||
BC026894.1 | - | 2 | 856 | |||
BC058709.1 | - | 14 | 3080 | 6841305 |