Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010554.4 | NP_034684.2 | 4, 5 | 379 | ||
GenBank mRNA | AK156449.1 | - | 4, 5 | 380 | ||
BC003727.1 | - | 3, 4 | 348 | 3599550 | ||
X01450.1 | - | 4, 5 | 379 |