Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il6ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010559.2 | NP_034689.2 | 5 | 897 | ||
GenBank mRNA | AK046337.1 | - | 5 | 897 | ||
AK082270.1 | - | 5 | 898 | |||
AK141197.1 | - | 5 | 932 | |||
BC019388.1 | - | 5 | 891 | |||
X51975.1 | - | 5 | 788 | |||
X51976.1 | - | 777 | ||||
X53802.1 | - | 5 | 897 |