Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epb4.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013513.2 | NP_038541.1 | 4 | 633 | ||
GenBank mRNA | AK133896.1 | - | 4 | 647 | ||
AK137484.1 | - | 4 | 714 | |||
AK143841.1 | - | 4 | 613 | |||
AK158219.1 | - | 4 | 551 | |||
AK158247.1 | - | 4 | 597 | |||
AK158370.1 | - | 4 | 560 | |||
AK165288.1 | - | 4 | 560 | |||
AK172410.1 | - | 4 | 597 | |||
BC053739.1 | - | 4 | 604 | |||
BC066194.1 | - | 4 | 633 | 30429971 | ||
BC066806.1 | - | 4 | 633 | 30429961 | ||
L35933.1 | - | 4 | 612 | |||
U03487.1 | - | 4 | 563 | |||
U04055.1 | - | 4 | 633 |