Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcer2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253737.1 | NP_001240666.1 | 7 | 462 | ||
NM_001253739.1 | NP_001240668.1 | 7 | 473 | |||
NM_001253743.1 | NP_001240672.1 | 8 | 522 | |||
NM_001253745.1 | NP_001240674.1 | 6 | 399 | |||
NM_001253746.1 | NP_001240675.1 | 6 | 399 | |||
NM_001253747.1 | NP_001240676.1 | 7 | 483 | |||
NM_013517.4 | NP_038545.1 | 8 | 603 | |||
GenBank mRNA | AK156654.1 | - | 8 | 606 | ||
AK172591.1 | - | 8 | 606 | |||
AY069980.1 | - | 6 | 365 | |||
AY069981.1 | - | 6 | 365 | |||
BC119164.1 | - | 7 | 470 | 8733913 | ||
BC119190.1 | - | 7 | 470 | 8733939 | ||
M34163.1 | - | 8 | 605 | |||
M99371.1 | - | 8 | 595 | |||
S67072.1 | - | 7 | 454 | |||
S67073.1 | - | 6 | 334 | |||
S67074.1 | - | 7 | 373 | |||
X64223.1 | - | 7 | 462 | |||
X64224.1 | - | 7 | 473 |