Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013531.4 | NP_038559.2 | 4 | 245 | ||
GenBank mRNA | AF277893.1 | - | 3 | 109 | ||
AF501886.1 | - | 3 | 109 | |||
AK036816.1 | - | 4 | 247 | |||
AK098139.1 | - | 4 | 178 | |||
AK152227.1 | - | 4 | 236 | |||
AK155309.1 | - | 4 | 304 | |||
AK158965.1 | - | 5 | 475 | |||
AK165084.1 | - | 4 | 236 | |||
AK167416.1 | - | 4 | 278 | |||
AK168302.1 | - | 4 | 201 | |||
AK169752.1 | - | 4 | 236 | |||
AK172147.1 | - | 5 | 378 | |||
BC028753.1 | - | 4 | 216 | 1348753 | ||
M63658.1 | - | 3 | 149 | |||
M87286.1 | - | 3 | 149 | |||
S86124.1 | - | 3 | 149 |