Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hgd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013547.3 | NP_038575.2 | 14 | 1596 | ||
GenBank mRNA | AK002584.1 | - | 13 | 1469 | ||
BC024369.1 | - | 13 | 1410 | 4974534 | ||
BC037628.1 | - | 14 | 1510 | 4973560 | ||
BC044758.2 | - | 13 | 1396 | 5099956 | ||
BC055029.1 | - | 14 | 1477 | 5054479 |