Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110251.1 | NP_001103721.1 | 14 | 1524 | ||
NM_013551.2 | NP_038579.2 | 14 | 1582 | |||
GenBank mRNA | AK143558.1 | - | 14 | 1449 | ||
AK153884.1 | - | 14 | 1512 | |||
AK166734.1 | - | 14 | 1480 | |||
AK167702.1 | - | 14 | 1541 | |||
AK199783.1 | - | 1 | 354 | |||
BC003861.1 | - | 14 | 1542 | 3487642 |