Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmmr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013552.2 | NP_038580.2 | 18 | 2617 | ||
GenBank mRNA | AF031932.1 | - | 18 | 2602 | ||
BC021427.1 | - | 17 | 2741 | 5035341 | ||
X64550.1 | - | 14 | 2230 |