Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099644.1 | NP_001093114.1 | 3 | 388 | ||
NM_013561.2 | NP_038589.2 | 3 | 388 | |||
GenBank mRNA | AF513221.1 | - | 3 | 265 | ||
AY605711.1 | - | 3 | 254 | |||
BC144799.1 | - | 3 | 310 | 9053347 | ||
BC145743.1 | - | 3 | 310 | 40131046 | ||
BY274793.1 | - | 3 | 389 | |||
M74425.1 | - | 3 | 311 | |||
X72395.1 | - | 3 | 272 |