Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013820.3 | NP_038848.1 | 18 | 3533 | ||
GenBank mRNA | AJ238540.1 | - | 18 | 3476 | ||
BC054472.1 | - | 18 | 3452 | 6465367 |