Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK088379.1 | - | 8 | 754 | ||
AK134525.1 | - | 5 | 409 | |||
AK163544.1 | - | 5 | 440 | |||
AV027352.2 | - | 2 | 192 | |||
BC010985.1 | - | 8 | 759 | 4038194 | ||
CX239267.1 | - | 8 | 752 | |||
M96265.1 | - | 8 | 786 |