Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK134525.1 | - | 8 | 859 | ||
AK163544.1 | - | 8 | 891 | |||
AV027352.2 | - | 5 | 643 | |||
M96265.1 | - | 10 | 1299 |