Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016697.3 | NP_057906.2 | 3 | 782 | ||
GenBank mRNA | AF185614.1 | - | 3 | 722 | ||
AK077942.1 | - | 3 | 760 | |||
AK146702.1 | - | 3 | 759 | |||
AK159394.1 | - | 3 | 751 | |||
AK159435.1 | - | 3 | 751 | |||
AK159764.1 | - | 3 | 759 | |||
AK168402.1 | - | 3 | 760 | |||
BC036126.1 | - | 3 | 729 | 4973409 |