Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159289.1 | NP_001152761.1 | 2 | 545 | ||
NM_001159290.1 | NP_001152762.1 | 2 | 200 | |||
NM_001242934.1 | NP_001229863.1 | 2 | 545 | |||
NM_016707.3 | NP_057916.1 | 2 | 545 | |||
GenBank mRNA | AF051525.1 | - | 2 | 483 | ||
AF169037.1 | - | 2 | 483 | |||
AF186018.1 | - | 2 | 426 | |||
AK004395.1 | - | 2 | 421 | |||
AK043677.1 | - | 2 | 344 | |||
AK045556.1 | - | 2 | 201 | |||
AK122557.1 | - | 2 | 454 | |||
AK140949.1 | - | 2 | 490 | |||
AK166352.1 | - | 2 | 546 | |||
BB649527.1 | - | 2 | 488 | |||
BC051418.1 | - | 2 | 333 | |||
BY735002.1 | - | 2 | 546 |