Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd17b10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016763.2 | NP_058043.3 | 3 | 217 | ||
GenBank mRNA | AF233685.1 | - | 3 | 217 | ||
AK002368.1 | - | 3 | 212 | |||
AK013340.1 | - | 3 | 213 | |||
AK077735.1 | - | 3 | 211 | |||
AK088685.1 | - | 3 | 233 | |||
BC027517.1 | - | 3 | 217 | 3376098 | ||
U96116.1 | - | 3 | 318 |