Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fanca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016925.3 | NP_058621.2 | 7 | 657 | ||
GenBank mRNA | AF178934.1 | - | 7 | 653 | ||
AF208116.1 | - | 7 | 659 | |||
AF208117.1 | - | 4 | 340 | |||
AF208118.1 | - | 7 | 659 | |||
AK084990.1 | - | 6 | 765 | |||
AK166112.1 | - | 7 | 642 | |||
BC150705.1 | - | 7 | 655 | 9087616 | ||
CF537383.1 | - | 7 | 574 |