Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016957.3 | NP_058653.1 | 5 | 382 | ||
GenBank mRNA | AK002374.1 | - | 5 | 298 | ||
AK078413.1 | - | 5 | 296 | |||
AK146285.1 | - | 5 | 353 | |||
AK146999.1 | - | 5 | 385 | |||
AK151383.1 | - | 5 | 298 | |||
AK152857.1 | - | 5 | 298 | |||
AK153881.1 | - | 298 | ||||
AK169050.1 | - | 5 | 298 | |||
BC083085.1 | - | 287 | 6516041 | |||
BC084680.1 | - | 5 | 234 | 6477991 | ||
BC090836.1 | - | 5 | 282 | 6819800 | ||
BC090837.1 | - | 5 | 272 | 6820123 | ||
BC092060.1 | - | 5 | 279 | 4020647 | ||
BC103792.1 | - | 282 | 6516212 | |||
CJ053675.1 | - | 5 | 384 | |||
X12944.1 | - | 239 |