Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289925.1 | NP_001276854.1 | 3 | 244 | ||
NM_001289927.1 | NP_001276856.1 | 2 | 118 | |||
NM_018781.3 | NP_061251.1 | 2 | 191 | |||
GenBank mRNA | AB221656.1 | - | 2 | 191 | ||
AF132128.1 | - | 2 | 191 | |||
AK141603.1 | - | 1 | 195 | |||
BC100734.1 | - | 2 | 190 | |||
BC103568.1 | - | 3 | 242 | 40045218 | ||
BC103569.1 | - | 2 | 186 | |||
BC103570.1 | - | 3 | 243 | 40045221 | ||
BY238697.1 | - | 2 | 119 | |||
S40835.1 | - | 2 | 99 |