Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289925.1 | NP_001276854.1 | 3 | 306 | ||
NM_001289927.1 | NP_001276856.1 | 2 | 180 | |||
NM_018781.3 | NP_061251.1 | 2 | 253 | |||
GenBank mRNA | AB221656.1 | - | 2 | 253 | ||
AF132128.1 | - | 2 | 253 | |||
AK141603.1 | - | 1 | 257 | |||
BC100734.1 | - | 2 | 252 | |||
BC103568.1 | - | 3 | 304 | 40045218 | ||
BC103569.1 | - | 2 | 248 | |||
BC103570.1 | - | 3 | 305 | 40045221 | ||
S40835.1 | - | 2 | 161 |