Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018869.3 | NP_061357.3 | 2, 3 | 979 | ||
GenBank mRNA | AF040746.1 | - | 2, 3 | 389 | ||
AK135076.1 | - | 2, 3 | 981 | |||
AK156465.1 | - | 2, 3 | 456 | |||
AK161821.1 | - | 2, 3 | 459 | |||
BC019379.1 | - | 2, 3 | 252 | 5116510 |