Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260504.1 | NP_001247433.1 | 6 | 738 | ||
NM_001260505.1 | NP_001247434.1 | 6 | 462 | |||
NM_031414.4 | NP_113602.2 | 6 | 462 | |||
NM_032944.3 | NP_116562.2 | 6 | 706 | |||
NR_048542.1 | 6 | 454 | ||||
XM_006715758.3 | XP_006715821.1 | 6 | 376 | |||
XM_011515449.2 | XP_011513751.2 | 6 | 388 | |||
XM_011515450.1 | XP_011513752.1 | 6 | 709 | |||
XM_011515452.2 | XP_011513754.2 | 6 | 389 | |||
GenBank mRNA | AF285599.1 | - | 6 | 358 | ||
AF332194.1 | - | 6 | 741 | |||
AK057271.1 | - | 6 | 410 | |||
AK292474.1 | - | 6 | 354 | |||
AK302689.1 | - | 6 | 354 | |||
BC033018.1 | - | 6 | 356 | |||
BC059374.1 | - | 6 | 369 | 5268051 | ||
DB065148.1 | - | 6 | 410 | |||
DB086591.1 | - | 6 | 465 | |||
DB089624.1 | - | 6 | 366 |