Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adgrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198894.1 | NP_001185823.1 | 7 | 1261 | ||
NM_018882.3 | NP_061370.2 | 7 | 1211 | |||
GenBank mRNA | AF166382.1 | - | 7 | 1261 | ||
AK087268.1 | - | 7 | 1013 | |||
AK133678.1 | - | 7 | 978 | |||
AK141886.1 | - | 7 | 1240 | |||
AK141894.1 | - | 7 | 1024 | |||
AK164776.1 | - | 7 | 1036 | |||
AK167547.1 | - | 6 | 965 | |||
BC034678.1 | - | 7 | 1007 | 3709247 |
Set Membership: |
G-protein coupled receptor |