Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fnbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038700.2 | NP_001033789.1 | 7 | 814 | ||
NM_001177648.1 | NP_001171119.1 | 7 | 814 | |||
NM_001177649.1 | NP_001171120.1 | 7 | 814 | |||
NM_001177650.1 | NP_001171121.1 | 5 | 511 | |||
NM_019406.3 | NP_062279.1 | 7 | 814 | |||
GenBank mRNA | AK052652.1 | - | 1 | 106 | ||
AK122308.1 | - | 7 | 814 | |||
AK143512.1 | - | 5 | 512 | |||
AK157135.1 | - | 7 | 805 | |||
AK170497.1 | - | 7 | 764 | |||
AK172100.1 | - | 7 | 758 | |||
BC003867.1 | - | 7 | 745 | 3490113 |