Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gria4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113180.1 | NP_001106651.1 | 8, 9 | 1537 | ||
NM_001113181.1 | NP_001106652.1 | 8, 9 | 1537 | |||
NM_019691.4 | NP_062665.3 | 8, 9 | 1537 | |||
GenBank mRNA | AB022913.1 | - | 8, 9 | 1271 | ||
AB079073.1 | - | 7, 8 | 1058 | |||
AB102777.1 | - | 7, 8 | 1058 | |||
AK031568.1 | - | 8, 9 | 1511 | |||
AK032103.1 | - | 8, 9 | 1538 | |||
AK135034.1 | - | 8, 9 | 1243 | |||
BC060697.1 | - | 8, 9 | 1445 | 6409918 | ||
CA320207.1 | - | 1, 2 | 157 |