Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fzd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021457.3 | NP_067432.2 | 1 | 2830 | ||
GenBank mRNA | AK143101.1 | - | 1 | 2650 | ||
BC053010.1 | - | 1 | 2585 | 5697795 |
Set Membership: |
G-protein coupled receptor |