Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163434.1 | NP_001156906.1 | 4, 5 | 653 | ||
NM_022310.3 | NP_071705.3 | 3, 4 | 735 | |||
GenBank mRNA | AJ002387.1 | - | 3, 4 | 514 | ||
AK004578.1 | - | 3, 4 | 730 | |||
AK076079.1 | - | 4, 5 | 649 | |||
AK077463.1 | - | 4, 5 | 653 | |||
AK144650.1 | - | 4, 5 | 649 | |||
AK144742.1 | - | 4, 5 | 649 | |||
AK146647.1 | - | 4, 5 | 652 | |||
AK148539.1 | - | 4, 5 | 652 | |||
AK149447.1 | - | 4, 5 | 652 | |||
AK151244.1 | - | 3, 4 | 602 | |||
AK151647.1 | - | 4, 5 | 651 | |||
AK151806.1 | - | 4, 5 | 652 | |||
AK152020.1 | - | 4, 5 | 649 | |||
AK152521.1 | - | 3, 4 | 732 | |||
AK152958.1 | - | 4, 5 | 652 | |||
AK159719.1 | - | 4, 5 | 649 | |||
AK160821.1 | - | 653 | ||||
AK166207.1 | - | 4, 5 | 649 | |||
AK166739.1 | - | 4, 5 | 649 | |||
AK167011.1 | - | 4, 5 | 649 | |||
AK168012.1 | - | 4, 5 | 653 | |||
AK168230.1 | - | 4, 5 | 648 | |||
AK169034.1 | - | 4, 5 | 653 | |||
AK192496.1 | - | 2, 3 | 244 | |||
AK195133.1 | - | 2, 3 | 209 | |||
AK197130.1 | - | 2, 3 | 200 | |||
AK200033.1 | - | 2, 3 | 207 | |||
AK219322.1 | - | 1, 2 | 114 | |||
BC005785.1 | - | 2, 3 | 369 | |||
BC050927.1 | - | 3, 4 | 712 | 6415084 | ||
D78645.1 | - | 3, 4 | 595 |