Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRSK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032430.1 | NP_115806.1 | 2 | 462 | ||
XM_005259327.3 | XP_005259384.1 | 2 | 527 | |||
GenBank mRNA | AF479826.1 | - | 4 | 360 | ||
AF479827.1 | - | 2 | 462 | |||
AL834275.1 | - | 2 | 85 | |||
AY458602.1 | - | 2 | 234 | |||
AY505124.1 | - | 2 | 185 | |||
HQ830199.1 | - | 2 | 207 |