Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301340.1 | NP_001288269.1 | 10 | 825 | ||
NM_001301341.1 | NP_001288270.1 | 12 | 952 | |||
NM_001301343.1 | NP_001288272.1 | 10 | 811 | |||
NM_001301344.1 | NP_001288273.1 | 9 | 753 | |||
NM_001301345.1 | NP_001288274.1 | 9 | 753 | |||
NM_025279.3 | NP_079555.1 | 10 | 825 | |||
GenBank mRNA | AK011428.1 | - | 10 | 830 | ||
AK051313.1 | - | 9 | 747 | |||
AK078777.1 | - | 12 | 953 | |||
AK088462.1 | - | 9 | 740 | |||
AK128891.1 | - | 10 | 818 | |||
AK135519.1 | - | 10 | 739 | |||
AK144245.1 | - | 8035 | ||||
AK150848.1 | - | 10 | 805 | |||
AK151689.1 | - | 9 | 707 | |||
AK152927.1 | - | 9 | 707 | |||
AK160615.1 | - | 10 | 795 | |||
AK160887.1 | - | 10 | 827 | |||
AK166656.1 | - | 10 | 830 | |||
AK166927.1 | - | 10 | 783 | |||
AK167600.1 | - | 10 | 816 | |||
AK168918.1 | - | 6 | 728 | |||
AK169064.1 | - | 10 | 815 | |||
AK197677.1 | - | 1 | 18 | |||
BC006694.1 | - | 10 | 792 | 3583037 | ||
BC089328.1 | - | 10 | 748 | 6402614 | ||
EU604547.1 | - | 8 | 606 |