Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifrd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025903.2 | NP_080179.1 | 3 | 407 | ||
GenBank mRNA | AK007687.1 | - | 3 | 408 | ||
AK149496.1 | - | 3 | 388 | |||
AK156625.1 | - | 3 | 401 | |||
AK165967.1 | - | 3 | 400 | |||
AK166130.1 | - | 3 | 475 | |||
AK166258.1 | - | 3 | 365 | |||
AY298860.1 | - | 3 | 236 | |||
BC023486.1 | - | 3 | 355 | |||
BC037434.1 | - | 3 | 363 | 2651172 |