Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026114.3 | NP_080390.1 | 8 | 1123 | ||
GenBank mRNA | AK003094.1 | - | 8 | 1441 | ||
AK009934.1 | - | 4 | 524 | |||
AK010592.1 | - | 8 | 1153 | |||
AK143499.1 | - | 8 | 1124 | |||
AK167763.1 | - | 8 | 1125 | |||
BC005463.1 | - | 8 | 1103 | 3591411 | ||
BC016448.1 | - | 8 | 1127 | 4234809 | ||
BC016497.1 | - | 8 | 1127 | 4501321 |