Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfxn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027324.5 | NP_081600.1 | 5 | 545 | ||
GenBank mRNA | AF325260.1 | - | 5 | 523 | ||
AK012650.1 | - | 5 | 544 | |||
AK151110.1 | - | 5 | 491 | |||
AK179717.1 | - | 4 | 330 | |||
BC005743.1 | - | 5 | 502 | 3599636 |