Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfxn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027324.5 | NP_081600.1 | 11 | 2486 | ||
GenBank mRNA | AF325260.1 | - | 11 | 2465 | ||
AK048238.1 | - | 1 | 46 | |||
AK076205.1 | - | 6 | 1855 | |||
AK084725.1 | - | 1 | 1426 |