Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301628.1 | NP_001288557.1 | 14 | 1933 | ||
NM_027604.4 | NP_081880.2 | 15 | 2020 | |||
GenBank mRNA | AF468037.1 | - | 15 | 1924 | ||
AK014891.1 | - | 9 | 1231 | |||
AK122301.1 | - | 14 | 1848 | |||
AK133852.1 | - | 14 | 1842 | |||
AK145749.1 | - | 15 | 1924 | |||
AK161469.1 | - | 15 | 1963 | |||
AK161957.1 | - | 14 | 1881 | |||
AK163315.1 | - | 14 | 1862 | |||
AK168755.1 | - | 14 | 1868 | |||
AK216706.1 | - | 1 | 23 | |||
BC003978.1 | - | 2 | 279 | |||
BC050042.1 | - | 15 | 1936 | 30022307 |