Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ide | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031156.3 | NP_112419.3 | 9, 10 | 1349 | ||
GenBank mRNA | AJ278422.1 | - | 9, 10 | 1369 | ||
AK004972.2 | - | 9, 10 | 1350 | |||
AK014703.1 | - | 1250 | ||||
AK029089.1 | - | 2, 3 | 145 | |||
AK128986.1 | - | 9, 10 | 1366 | |||
BC041675.1 | - | 9, 10 | 1381 | 4951971 |