Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK098074.1 | - | 2 | 196 | ||
AK157831.1 | - | 2 | 197 | |||
AK167845.1 | - | 2 | 200 | |||
AK167910.1 | - | 2 | 200 | |||
AK169594.1 | - | 2 | 196 | |||
BC006722.1 | - | 2 | 221 | 3711399 | ||
BC094900.1 | - | 2 | 170 | 30063483 | ||
U27129.1 | - | 2 | 148 |