Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ier3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133662.2 | NP_598423.1 | 2 | 1075 | ||
GenBank mRNA | AK003466.1 | - | 2 | 1101 | ||
AK051003.1 | - | 2 | 1105 | |||
AK170477.1 | - | 2 | 1105 | |||
BC006950.1 | - | 2 | 1076 | 2811734 | ||
X67644.1 | - | 1889 |