Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il13ra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080147.1 | - | 4 | 453 | ||
AK082889.1 | - | 4 | 448 | |||
AK089984.1 | - | 4 | 484 | |||
AK143292.1 | - | 4 | 453 | |||
AK154675.1 | - | 4 | 508 | |||
BC052425.1 | - | 4 | 476 | 6835500 | ||
BC059939.1 | - | 4 | 444 | 6477224 | ||
S80963.1 | - | 4 | 481 |