Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144512.2 | NP_653095.1 | 11 | 1294 | ||
GenBank mRNA | AK149557.1 | - | 11 | 1302 | ||
BC021606.1 | - | 2 | 152 | |||
BC023117.1 | - | 11 | 1282 | 4240641 | ||
BC029637.1 | - | 11 | 1294 | 4195373 |