Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grik1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010348.3 | NP_034478.1 | 10 | 1907 | ||
NM_146072.4 | NP_666184.2 | 10 | 1907 | |||
GenBank mRNA | AK042635.1 | - | 10 | 1908 | ||
AK043542.1 | - | 10 | 1887 | |||
AK043737.1 | - | 10 | 1871 | |||
AK044250.1 | - | 10 | 1866 | |||
AK044420.1 | - | 10 | 1862 | |||
AK046122.1 | - | 11 | 1959 | |||
AK048634.1 | - | 10 | 1878 | |||
AK133667.1 | - | 10 | 1893 | |||
AK215131.1 | - | 1 | 20 | |||
BC031822.1 | - | 9 | 1329 | 4501004 | ||
BC049275.1 | - | 9 | 1329 | 4501004 | ||
X66118.1 | - | 10 | 1893 |