Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THOC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005131.2 | NP_005122.2 | 11 | 925 | ||
XM_011525772.2 | XP_011524074.1 | 11 | 925 | |||
XM_011525773.1 | XP_011524075.1 | 12 | 955 | |||
XM_017026103.1 | XP_016881592.1 | 8 | 2445 | |||
XM_017026104.1 | XP_016881593.1 | 11 | 925 | |||
GenBank mRNA | AK225067.1 | - | 11 | 901 | ||
AK225709.1 | - | 11 | 889 | |||
AK314755.1 | - | 11 | 891 | |||
AY573303.1 | - | 11 | 885 | |||
BC010381.1 | - | 11 | 916 | 4046908 | ||
L36529.1 | - | 11 | 899 |