Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs3st3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178870.5 | NP_849201.1 | 2 | 2330 | ||
GenBank mRNA | AK032282.1 | - | 2 | 1298 | ||
AK051284.1 | - | 2 | 2331 | |||
BC094320.1 | - | 2 | 2384 | 6400013 |