Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP4R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042388.2 | NP_001035847.1 | 13, 14 | 1957 | ||
NM_005134.3 | NP_005125.1 | 13, 14 | 1906 | |||
NR_052003.1 | 13, 14 | 1924 | ||||
XM_011525775.2 | XP_011524077.1 | 13, 14 | 2083 | |||
XM_011525776.1 | XP_011524078.1 | 13, 14 | 1947 | |||
XM_011525779.2 | XP_011524081.1 | 10, 11 | 1750 | |||
XM_017026105.1 | XP_016881594.1 | 12, 13 | 1924 | |||
XM_017026106.1 | XP_016881595.1 | 12, 13 | 1761 | |||
XM_017026107.1 | XP_016881596.1 | 12, 13 | 1776 | |||
GenBank mRNA | AF100744.1 | - | 2, 3 | 124 | ||
AF111106.1 | - | 13, 14 | 1876 | |||
AF200478.1 | - | 13, 14 | 1856 | |||
AK027064.1 | - | 13, 14 | 1908 | |||
AK292538.1 | - | 13, 14 | 1837 | |||
AK293869.1 | - | 13, 14 | 1821 | |||
AK295533.1 | - | 4, 5 | 691 | |||
AK315106.1 | - | 13, 14 | 1864 | |||
BC060829.1 | - | 13, 14 | 1858 | 30342594 |