Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243043.1 | NP_001229972.1 | 23 | 3273 | ||
NM_001243044.1 | NP_001229973.1 | 22 | 3055 | |||
NM_007454.3 | NP_031480.2 | 21 | 3046 | |||
GenBank mRNA | AK034140.1 | - | 23 | 3276 | ||
AK155892.1 | - | 21 | 2966 | |||
AK159273.1 | - | 21 | 2937 | |||
AK165747.1 | - | 22 | 2974 | |||
AK170376.1 | - | 21 | 2957 | |||
BC008513.1 | - | 21 | 2859 | 3601435 | ||
BC066827.1 | - | 12 | 1590 | |||
Y07919.1 | - | 21 | 2901 |