Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007457.2 | NP_031483.1 | 4 | 430 | ||
GenBank mRNA | AK002225.1 | - | 4 | 408 | ||
BC052692.1 | - | 4 | 434 | 5714625 | ||
M62418.1 | - | 4 | 430 |