Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077264.1 | NP_001070732.1 | 23 | 3353 | ||
NM_007458.2 | NP_031484.1 | 24 | 3419 | |||
GenBank mRNA | AK135424.1 | - | 23 | 3282 | ||
BC031433.1 | - | 24 | 3369 | 4481643 |
Gene Symbol: Fuz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CO041869.1 | - | 1 | 245 |